Learn More
Buerkle™ Squeeze-Fix Hose Clamp
Product Code. 10253652 Shop All Bürkle Products
Click to view available options
Tubing Size:
1 to 5 mm
3 to 10 mm
6 to 15 mm
Overall Length (Metric):
25 mm
40 mm
55 mm
Unit Size:
Each
Product Code. 10253652
Brand: Buerkle™ 86180003
Only null left
Add to basket
Add to basket
Using these clips, hoses can be compressed partially or completely as required. Buerkle™ Squeeze-Fix Hose Clamps are made from POM, do not rust, and can be operated with one hand. Permanently elastic, hygienic and sterilizable.
Using these clips, hoses can be compressed partially or completely as required.
  • Can be operated with one hand
  • Made from polyoxymethylene plastic (POM)
  • Does not rust
  • Permanently elastic and hygienic
  • White
  • Sterilizable
TRUSTED_SUSTAINABILITY
Specifications
Material POM
Overall Length (Metric) 55 mm
For Use With Hoses
Tubing Size 6 to 15 mm
Color White
Product Type Hose Clamp