Learn More
Squalene 2,3:22,23-Dioxide, TRC
Product Code. 30134744
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134744 250 mg 250mg
30011130 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30134744 Supplier Toronto Research Chemicals Supplier No. TRCS683810250MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 31063-19-1
Molecular Formula C30H50O2
Molecular Weight (g/mol) 442.72
Synonym 2,3:22,23-Dioxidosqualene, Squalene 2,3:22,23-Diepoxide, (all-E)-2,3:22,23-Diepoxy-2,6,10,15,19,23-hexamethyl-6,10,14,18-tetracosatetraene, 2,2'-(3,7,12,16-Tetramethyl-3,7,11,15-octadecatetraene-1,18-diyl)bis[3,3-dimethyl-oxirane]
IUPAC Name 3-[(3E,7E,11E,15E)-18-(3,3-dimethyloxiran-2-yl)-3,7,12,16-tetramethyloctadeca-3,7,11,15-tetraenyl]-2,2-dimethyloxirane
SMILES C/C(CCC1C(O1)(C)C)=C\CC/C(C)=C/CC/C=C(C)/CC/C=C(C)/CCC2C(O2)(C)C
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C30H50O2/c1-23(15-11-17-25(3)19-21-27-29(5,6)31-27)13-9-10-14-24(2)16-12-18-26(4)20-22-28-30(7,8)32-28/h13-14,17-18,27-28H,9-12,15-16,19-22H2,1-8H3/b23-13+,24-14+,25-17+,26-18+
Formula Weight 442.3811
Percent Purity >95%
Chemical Name or Material Squalene 2,3:22,23-Dioxide
Grade HPLC