Learn More
Trajan SPure LS H2O/HC Filter 1/4 SS Compression Fittings
Brand: Trajan 202325L-SS
SPure LS H2O/HC Filter 1/4 SS Compression Fittings
Safety and Handling
- SPure LS H2O/HC Filter 1/4 SS Compression Fittings
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1A
Hazard Statement
- H350-May cause cancer.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contain: carbon, Quartz
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.