Learn More
- Home
- Products
- Tubing
- Plastic Tubing
- Tygon SPT-3350 Platinum-Cured Silicone Tubing
Tygon SPT-3350 Platinum-Cured Silicone Tubing
Product Code. 30053015 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1 in.
1-1/2 in.
1-1/4 in.
1/16 in.
1/2 in.
1/32 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
7/16 in.
Diameter (English) Outer:
1 in.
1-1/2 in.
1-1/4 in.
1/2 in.
1/4 in.
1/8 in.
11/16 in.
13/16 in.
2 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
7/16 in.
7/32 in.
7/8 in.
9/16 in.
Unit Size:
15m
| Product Code. | Diameter (English) Inner | Diameter (English) Outer | unitSize |
|---|---|---|---|
| 30053015 | 1/16 in. | 3/16 in. | 15m |
| 30053509 | 1/4 in. | 3/8 in. | 15m |
| 30053959 | 3/8 in. | 1/2 in. | 15m |
| 30053098 | 3/16 in. | 5/16 in. | 15m |
| 30054135 | 3/16 in. | 7/16 in. | 15m |
| 30053030 | 1/16 in. | 1/8 in. | 15m |
| 30053420 | 3/32 in. | 5/32 in. | 15m |
| 30053915 | 7/16 in. | 9/16 in. | 15m |
| 30053674 | 3/8 in. | 5/8 in. | 15m |
| 30053763 | 1/4 in. | 5/16 in. | 15m |
| 30053277 | 3/4 in. | 1 in. | 15m |
| 30053357 | 3/8 in. | 9/16 in. | 15m |
| 30053193 | 1/2 in. | 3/4 in. | 15m |
| 30053544 | 5/8 in. | 7/8 in. | 15m |
| 30053721 | 1 in. | 1-1/4 in. | 15m |
| 30054107 | 1/2 in. | 5/8 in. | 15m |
| 30053476 | 7/16 in. | 5/8 in. | 15m |
| 30054184 | 3/16 in. | 3/8 in. | 15m |
| 30053526 | 1/2 in. | 11/16 in. | 15m |
| 30053711 | 1-1/2 in. | 2 in. | 15m |
| 30054333 | 5/32 in. | 7/32 in. | 15m |
| 30053671 | 1/32 in. | 3/32 in. | 15m |
| 30053825 | 1-1/4 in. | 1-1/2 in. | 15m |
| 30054156 | 3/32 in. | 7/32 in. | 15m |
| 30053351 | 1/4 in. | 1/2 in. | 15m |
| 30053768 | 3/16 in. | 1/4 in. | 15m |
| 30053003 | 5/8 in. | 13/16 in. | 15m |
| 30053126 | 5/16 in. | 7/16 in. | 15m |
| 30053099 | 5/16 in. | 1/2 in. | 15m |
| 30053335 | 1/4 in. | 7/16 in. | 15m |
| 30053823 | 1/8 in. | 1/4 in. | 15m |
| 30053559 | 1/8 in. | 3/16 in. | 15m |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
Ideal for food and beverage transfer. Minimal e in., tractable maintains fluid integrity. Up to three times smoother than other silicone tubing. FDA 21 CFR 175.300 compliant. Free technical application support available to assist with tubing selection
Specifications
Specifications
| Certifications/Compliance | USP;NSF-51;FDA |
| Diameter (English) Inner | 1/16 in. |
| Material | Tygon 3350 |
| Diameter (English) Outer | 3/16 in. |
| Overall Length (English) | 50 ft. |
| Sterilization Method | Yes |
| Color | Translucent |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.