Learn More
Spreader Blue Polystyrene Wedge-Shaped Spreader
Product Code. 12342048
Click to view available options
Quantity:
100 bags of 5
25 Bags of 20
Unit Size:
Pack of 500
Product Code. 12342048
Brand: Microspec D6
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
High impact wedge-shaped disposable bacterial spreaders
High impact polystyrene disposable bacterial spreaders. Wedge shaped to reduce tendency to flip, or traditional “L” shape. Packs of 500 contain re-sealable inner bags of 5 sterile spreaders.
Specifications
Disposable Single-use
Color Blue
Quantity 100 bags of 5
Shape Wedge-shaped
Material Polystyrene
Product Type Spreader
For Use With (Application) High impact Polystyrene disposable bacterial spreaders