Learn More
Spreader Blue Polystyrene L-Shaped Spreader
Product Code. p-8000346
Click to view available options
Quantity:
100 Bags of 5
25 bags of 20
Unit Size:
Pack of 500
Product Code. 12322048
Brand: Microspec E6
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
High impact L-shape disposable bacterial spreaders
High impact polystyrene disposable bacterial spreaders. Wedge shaped to reduce tendency to flip, or traditional “L” shape. Packs of 500 contain re-sealable inner bags of either 20 or 5 sterile spreaders.
Specifications
Disposable Single-use
Color Blue
Quantity 100 Bags of 5
Shape L-Shape
Material Polystyrene
Product Type Spreader
Sterility Sterile
For Use With (Application) High impact Polystyrene disposable bacterial spreaders