Learn More
uvex 9193 Sportstyle Safety Spectacles
Product Code. 17787689 Shop All Uvex Products
Ansicht ändern
Click to view available options
Supravision ETC, Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
17787689 Supravision ETC, Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17787689 Lieferant uvex Lieferanten-Nr. 9193415
  • ONLINE EXCLUSIVE PRICE
    14.05€
    13.21€ / Pack of 1
    Save 0.84€
    5% Off
Alle Werbeaktionen anzeigen
Estimated Shipment: 28-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Innovative safety spectacles featuring a sporty design, combined with maximum functionality and safety. Feather-light construction weighs only 23 grams, making these spectacles comfortable to wear for extended periods of time.
Modern eye protection. The uvex sportstyle stands for a new generation of innovative safety glasses: a clean, sporty design combining maximum functionality and protection – these lightweight safety glasses are a joy for everyone.
General features
  • Sporty, lightweight safety glasses with contoured lens for a great fit on narrow or wider faces
  • Robust polycarbonate lens with proven uvex supravision coating technology – permanently anti-fog and scratch-resistant lens
  • Frame color: anthracite, blue
  • PC lens: clear
Protection features
  • Certified in line with EN 166 (personal eye protection) and EN 170 (UV filters)
  • Marking: W 166 FT CE – 2C-1.2 W1 FTKN CE
  • UV400 – 100% protection against dangerous UVA and UVB radiation to 400 nm
Comfort features
  • The combination of soft padding around the forehead and ergonomically shaped arms with soft ear pieces ensures a comfortable and secure fit without creating pressure points
  • Soft, adjustable nose piece makes it possible to adjust the glasses to the wearer and ensures a comfortable, non-slip, pressure-free fit
TRUSTED_SUSTAINABILITY
Spezifikation
Lens Type Supravision ETC
Lens Coating Anti-fog
Color Anthracite, Blue
Certifications/Compliance (Europe) EN 166, EN 170
Lens Tint Clear
Product Type Ergonomic Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Standard
Mehr anzeigen Weniger anzeigen