Learn More
Carbolite™ Split Tube Furnace
Product Code. 12113360
Click to view available options
Length (Metric):
350mm
700mm
Wattage:
1000 w
3000 w
Unit Size:
Each
Product Code. 12113360
Brand: Carbolite™ VST120200230SN+&023216P1
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Analysis Time 45 min.
Type Split Tube Furnace
Temperature Stability 1200°C
Dimensions (L x W x H) Exterior 350 W x 300 H mm
Length (Metric) 350mm
Voltage 220 V
Wattage 1000 w