Learn More
DWK Life Sciences™ Spirit Lamp, from Soda-lime Glass, without filling tubulature, with ground over-cap, with socket and wick
Product Code. 15201859 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15201859 100 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15201859 Supplier DWK Life Sciences™ Supplier No. 234002455
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The spirit lamp is an alternative to the gas-powered Bunsen burner and is also suitable for heating cooking glasses and the materials they contain.
In order for substances to react with each other, a low heat input is often required. The spirit burner is used classically and in areas where direct gas extraction is not possible. It is an alternative to the well-known Bunsen burner.
  • Without filling tubulature
  • The ground cap ensures a tight seal and quick suffocation of the flame
TRUSTED_SUSTAINABILITY
Specifications
Material Soda Lime Glass
Capacity (Metric) 100 mL
Description With socket and wick
Diameter (Metric) 75 mm
For Use With (Application) Heating; Use with Spritus
Height (Metric) 103 mm
No. per Pack 10 Pk.