missing translation for 'onlineSavingsMsg'
Learn More

Spiramycin Adipate, TRC

Product Code. 30410069
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410069 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410069 Supplier Toronto Research Chemicals Supplier No. TRCS682365250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Spiramycin adipate base
CAS 68880-55-7
Molecular Formula C43H74N2O14 C6H10O4
Quantity 250 mg
Synonym Hexanedioic Acid','Compd. With Spiramycin, Mamivert, Spiramycin Adipate, Spiramycin','Hexanedioate (salt), Suanovil
InChI Formula InChI=1S/C43H74N2O14.C6H10O4/c1-24-21-29(19-20-46)39(59-42-37(49)36(45(9)10)38(27(4)56-42)58-35-23-43(6,51)41(50)28(5)55-35)40(52-11)31(47)22-33(48)53-25(2)15-13-12-14-16-32(24)57-34-18-17-30(44(7)8)26(3)54-34;7-5(8)3-1-2-4-6(9)10/h12-14,16,20,24-32,34-42,47,49-51H,15,17-19,21-23H2,1-11H3;1-4H2,(H,7,8)(H,9,10)/b13-12+,16-14+;/t24?,25?,26?,27?,28?,29-,30-,31+,32-,34?,35-,36+,37+,38+,39-,40?,41-,42-,43?;/m0./s1
SMILES O=C1O[C@H](C)C/C=C/C=C/[C@H](OC2CC[C@H](N(C)C)[C@@H](C)O2)[C@H](C)C[C@H](CC=O)[C@H](O[C@H]3[C@H](O)[C@@H](N(C)C)[C@H](O[C@@H]4O[C@@H](C)[C@H](O)[C@@](O)(C)C4)[C@@H](C)O3)[C@@H](OC)[C@H](O)C1.OC(CCCCC(O)=O)=O
IUPAC Name 2-((4R,5S,6S,7R,9R,10R,11E,13E,16R)-6-(((2S,3R,4R,5S,6R)-5-(((2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-4-(dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-10-(((5S,6R)-5-(dimethylamino)-6-methyltetrahydro-2H-pyran-2-yl)oxy)-4-hydroxy-5-methoxy-9,16-dimethyl-2-oxooxacyclohexadeca-11,13-dien-7-yl)acetaldehyde adipate
Molecular Weight (g/mol) 988.57
Formula Weight 988.57
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.