missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spiramycin Adipate, TRC
Specifications
Specifications
| Chemical Name or Material | Spiramycin adipate base |
| CAS | 68880-55-7 |
| Molecular Formula | C43H74N2O14 C6H10O4 |
| Quantity | 1 g |
| Synonym | Hexanedioic Acid','Compd. With Spiramycin, Mamivert, Spiramycin Adipate, Spiramycin','Hexanedioate (salt), Suanovil |
| InChI Formula | InChI=1S/C43H74N2O14.C6H10O4/c1-24-21-29(19-20-46)39(59-42-37(49)36(45(9)10)38(27(4)56-42)58-35-23-43(6,51)41(50)28(5)55-35)40(52-11)31(47)22-33(48)53-25(2)15-13-12-14-16-32(24)57-34-18-17-30(44(7)8)26(3)54-34;7-5(8)3-1-2-4-6(9)10/h12-14,16,20,24-32,34-42,47,49-51H,15,17-19,21-23H2,1-11H3;1-4H2,(H,7,8)(H,9,10)/b13-12+,16-14+;/t24?,25?,26?,27?,28?,29-,30-,31+,32-,34?,35-,36+,37+,38+,39-,40?,41-,42-,43?;/m0./s1 |
| SMILES | O=C1O[C@H](C)C/C=C/C=C/[C@H](OC2CC[C@H](N(C)C)[C@@H](C)O2)[C@H](C)C[C@H](CC=O)[C@H](O[C@H]3[C@H](O)[C@@H](N(C)C)[C@H](O[C@@H]4O[C@@H](C)[C@H](O)[C@@](O)(C)C4)[C@@H](C)O3)[C@@H](OC)[C@H](O)C1.OC(CCCCC(O)=O)=O |
| IUPAC Name | 2-((4R,5S,6S,7R,9R,10R,11E,13E,16R)-6-(((2S,3R,4R,5S,6R)-5-(((2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-4-(dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-10-(((5S,6R)-5-(dimethylamino)-6-methyltetrahydro-2H-pyran-2-yl)oxy)-4-hydroxy-5-methoxy-9,16-dimethyl-2-oxooxacyclohexadeca-11,13-dien-7-yl)acetaldehyde adipate |
| Molecular Weight (g/mol) | 988.57 |
| Formula Weight | 988.57 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?