Learn More
Buerkle™ Spiral Screw for PowderProof™ Sampler
Product Code. 11887383 Shop All Bürkle Products
Click to view available options
Length (Metric):
90 cm
Unit Size:
Each
Product Code. 11887383
Brand: Buerkle™ 53020912
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
PowderProof is the practically proven professional solution for mobile sampling of powders and granules. Different lances and augers can be attached to the robust drive unit and swapped without difficulty. Stainless steel V4A (1.4404)
PowderProof is the practically proven professional solution for mobile sampling of powders and granules. Different lances and augers can be attached to the robust drive unit and swapped without difficulty. Stainless steel V4A (1.4404)
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (V4A)
For Use With (Equipment) For up to 2 mm dia. Powders
Length (Metric) 90 cm