missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SPINK1 (Human) Recombinant Protein
Human SPINK1 full-length ORF ( NP_003113.2, 1 a.a. - 79 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00006690-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MKVTGIFLLSALALLSLSGNTGADSLGREAKCYNELNGCTKIYDPVCGTDGNTYPNECVLCFENRKRQTSILIQKSGPC
Specifications
NP_003113.2 | |
serine peptidase inhibitor, Kazal type 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SPINK1 | |
GST |
6690 | |
Wheat germ expression system | |
25 μg | |
PCTT, PSTI, Spink3, TATI | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |