Learn More
Justrite™ SpillSlope™ Steel Shelf for 22 Gallon Undercounter Safety Cabinet
Description
- Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump. They adjust on 3 in. (76-mm) centers for versatile storage
- Fits 22 gal. Undercounter cabinet
Specifications
Specifications
| For Use With (Equipment) | 22 gal. Undercounter Cabinet |
| Depth (English) Exterior | 18 in. |
| Material | Steel |
| Width (English) Exterior | 31.56 in. |