Learn More
Cole Parmer Spill-X Refill
Product Code. 13141176 Shop All Cole-Parmer Products
Change view
Click to view available options
For Use With (Application):
Acid Agent Spill
Color:
Red
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Color Unit Size
13141176 Acid Agent Spill Red Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13141176 Supplier Cole Parmer Supplier No. 8629250
  • ONLINE EXCLUSIVE PRICE
    317.00€
    244.20€ / Pack of 6
    Save 72.80€
    22% Off
 Restricted Item
Estimated Shipment: 06-01-2027
Cole Parmer Spill-X Refill is an easy-to-store carton of six handy polypropylene containers to replace specific or multipurpose Spill Treatment Kit containers.
Specifications
Color Red
Material Polypropylene
Type Spill Treatment Refill
For Use With (Application) Acid Agent Spill
For Use With (Equipment) Spill Treatment Kit Containers
Model Spill-X Refill
Packaging Quantity 6/Cs.
compliance-icons
Product Identifier
  • X6 SPILL-X-A ACID AGENT REFILL,
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: magnesium oxide, fullers earth, sodium carbonate