missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ SPC™ Absorbents Spill Containment Berm, QuickBerm Rigid Lock
Click to view available options
Capacity (Metric) Sump:
662 L
890 L
2820 L
Unit Size:
Each
Description
The extreme durable Rigid-Lock QuickBerm offers immediate, secure deployment and spill-readiness.
- The built-in Rigid-Lock wall supports maximise usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter.
- The high performance material withstands a broad range of chemicals including petroleum, oils, grease, and most acids and a broad UV resistance - durable enough for extended wear.
Specifications
Specifications
| Capacity (English) Sump | 235.113 gal. |
| Capacity (Metric) Sump | 890 L |
| Color | Black |
| Material | PVC-coated Fabric |
| Type | Containment Berm |
| For Use With (Application) | Beverage Processing, Chemical, Food Processing, Government, Healthcare, Manufacturing, Maritime, Mass transit, Oil and Gas, Pharmaceutical, Process, Warehouse and Logistics |
| Model | Spill Containment |
| Length (English) | 8.661 ft. |
| Length (Metric) | 2.64 m |
| Width (English) | 4.665 ft. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction