Learn More
Brady™ Spill Barrier
Product Code. 16368877 Shop All Brady Products
Change view
Click to view available options
Color:
Yellow
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16368877 Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16368877 Supplier Brady™ Supplier No. 813921
  • ONLINE EXCLUSIVE PRICE
    765.40€
    616.51€ / Pack of 1
    Save 148.89€
    19% Off
In Stock
SB-3 is a spill barrier, made from chemical-resistant polyurethane that is used to contain or divert spills easily and quickly.
  • Resistant to water, oil and many chemicals
  • Thanks to the interlocking ends no additional connectors are needed to connect multiple barriers
  • Yellow color for high visibility
  • Re-usable after cleaning with soap and water
  • Works best on smooth surfaces
  • Ideal as a barrier in doorways during washdown operations, as a dam to control or contain spilled liquids or as a diverter to direct liquids away from valuable equipment or vulnerable areas
Specifications
Color Yellow
For Use With (Application) Containing/Diverting Spills
Dimensions (L x W x H) 3 m x 100 mm x 9 cm
Alternate Part Number SB-3