Learn More
R&D Systems™ Cultrex Spheroid Invasion Extracellular Matrix
Click to view available options
Quantity:
6 mL
missing translation for 'unitSize'
6mL
Description
- Dispense into working aliquots if needed; repeated freeze-thaws will destroy product integrity.
- Source: Murine Engelbreth-Holm-Swarm (EHS) tumor and bovine extensor tendon.
Specifications
Specifications
| Content And Storage | Product is stable for a minimum of 3 months from date of shipment. Store at -80° C; keep frozen. Dispense into working aliquots if needed; repeated freeze-thaws will destroy product integrity. |
| Quantity | 6 mL |
| Stability | Product is stable for a minimum of 3 months from date of shipment. |