Learn More
KGW Isotherm™ Spherical Dewar Flasks
Product Code. p-8290033
Click to view available options
Capacity (Metric):
3 L
5 L
Unit Size:
Each
Product Code. 10145500
Brand: KGW Isotherm™ 1292
Only null left
Add to basket
Add to basket
For the storage and transport of liquid nitrogen. KGW Isotherm™ Spherical Dewar Flasks are made from borosilicate glass 3.3.
  • Low evaporation rate
  • Good tipping stability
  • Fully silvered as standard
  • Brushed aluminum cover
  • Includes handle and loose lied lid
Specifications
Cap Lid Type Loose
Color Silver
Depth (Metric) Inside 305mm
Diameter (Metric) Outer 225mm
For Use With (Application) Laboratory technique, Medicinal technique
Height (Metric) Inner 210mm
Height (Metric) 380mm
Includes Lid Handle
Base Material Borosilicate Glass
Capacity (Metric) 3 L
Diameter (Metric) Inner 205 mm
Material Aluminum