Learn More
General Tools Digital Manometer
Product Code. 15200748 Shop All Cole-Parmer Products
Change view
Click to view available options
Pressure Range:
100 psi
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pressure Range Unit Size
15200748 100 psi Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15200748 Supplier General Tools Supplier No. 6860306
  • ONLINE EXCLUSIVE PRICE
    461.00€
    372.82€ / Pack of 1
    Save 88.18€
    19% Off
Estimated Shipment: 09-12-2026
Use the Digital Manometer to read gauge and differential pressure in 11 units of measure.
  • Ideal for measuring low differential pressure in clean rooms, test and balance, HVAC, pneumatic systems, and computer peripherals.
  • Large backlight LCD, maximum, minimum, hold, and auto power off functions.
  • Built-in RS-232 port downloads information to your PC.
  • These manometers are for use with noncorrosive gases and air only, not liquids.
Specifications
Certifications/Compliance CE Compliance
For Use With (Application) Ideal for measuring low differential pressure in clean rooms, test and balance, HVAC, pneumatic systems, and computer peripherals
Includes One 9 Volt Battery
Size (English) Fitting 0.2 in. Barb
Size (Metric) Fitting 4.57 mm Barb
Width (English) 9.75 in.
Width (Metric) 247.65 mm
Accuracy ±0.3% Full-scale
Display Type LCD
Pressure Range 100 psi
Product Type Digital Manometer