Learn More
3M™ Speedglas™ Outside Protection Plate
Product Code. 17859306 Shop All 3M Products
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17859306 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17859306 Supplier 3M™ Supplier No. 7100092252
  • ONLINE EXCLUSIVE PRICE
    413.00€
    273.33€ / Pack of 10
    Save 139.67€
    33% Off
Estimated Shipment: 15-12-2026
3M™ Speedglas™ Outside Protection Plates are designed for use with 3M™ Speedglas™ Welding Helmets and Filters and protect the welding filter from sparks and spatter.
  • 3M™ Speedglas™ Outside Protection Plates are a replacement part designed to be used when the original has been damaged or worn out.
  • Help protect your welding filter from sparks and spatter by ensuring that your Outside Protection Plate is functioning properly.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z89.1-2010
Color Clear
For Use With (Equipment) 9002NC Welding Helmets, 9100MP Welding Helmets, G5-02 Welding Helmets
Product Type Outside Protection Plate
Quantity 10/Pk.
Size Standard