Learn More
3M™ Speedglas™ Outer Protection Plate
Description
- Replacement outer protection plate for use with 3M™ Speedglas™ Welding Helmet 100 Series
- Complies with the harmonized European Standards EN 166, class 1BT
- Protects the welding filter from sparks and spatter
- Clean regularly with a soft cloth and replace if pits or scratches occur
- Check your equipment regularly to increase service life
Specifications
Specifications
| Certifications/Compliance | EN166:BT, EN 166 |
| For Use With (Equipment) | 3M™ Speedglas™ Welding Helmet Series 100 |
| Material | Polycarbonate |
| Product Type | Outer Protection Plate |