Learn More
3M™ Speedglas™ Outer Protection Plate
Product Code. 15307389
Change view
Click to view available options
:
Model: 776000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15307389 Model: 776000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15307389 Supplier 3M™ Supplier No. 7000000233
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
3M™ Outer Protection Plate protects the welding filter from sparks and spatter.
  • Replacement outer protection plate for use with 3M™ Speedglas™ Welding Helmet 100 Series
  • Complies with the harmonized European Standards EN 166, class 1BT
  • Protects the welding filter from sparks and spatter
  • Clean regularly with a soft cloth and replace if pits or scratches occur
  • Check your equipment regularly to increase service life
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN166:BT, EN 166
For Use With (Equipment) 3M™ Speedglas™ Welding Helmet Series 100
Material Polycarbonate
Product Type Outer Protection Plate