Learn More
3M™ Speedglas™ Inner Cover Plate 9002X
Product Code. 15709637
Change view
Click to view available options
For Use With (Application):
Increase 1 shade
Increase 2 shades
Protection
Unit Size:
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) unitSize
15709637 Protection Pack of 5
15760098 Increase 1 shade Pack of 5
15710098 Increase 2 shades Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15709637 Supplier 3M™ Supplier No. 7000000184
  • ONLINE EXCLUSIVE PRICE
    26.95€
    21.35€ / Pack of 5
    Save 5.60€
    20% Off
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
3M™ Speedglas™ Inner Cover Plate 9002X is designed to protect the inside surface of the filter from spark and spatter.
  • The inner cover plate is designed to protect the inside surface of the filter from spark and spatter
  • Replace anytime the plate becomes deeply pitted, scratched or too dirty to clean with soft cloth or lens paper
  • Regularly clean with a soft cloth and replace if any pits or scratches occur
  • Can be used to increase +1 or +2 dark shades of the welding filter
  • Failure to use an inside protection plate with the filter voids the warranty if spark or spatter cause damage to the glass
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Dimensions (L x W) 53 x 103 cm
For Use With (Equipment) 3M Speedglas Welding Filter
For Use With (Application) Protection
Product Type Inner Cover Plate