Learn More
3M™ Speedglas™ Heat Reflecting Silver Front Panel
Product Code. 15879634 Shop All 3M Products
Change view
Click to view available options
Color: Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15879634 Color: Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15879634 Supplier 3M™ Supplier No. 7000028045
  • ONLINE EXCLUSIVE PRICE
    42.00€
    30.97€ / Pack of 1
    Save 11.03€
    26% Off
Estimated Shipment: 20-01-2027
3M™ Speedglas™ Heat Reflecting Silver Front Panel for use with 3M™ Speedglas™ Welding Helmets 9100/9100 Air and 9100-QR.
  • For maximum safety and comfort, make it a habit to check your equipment regularly
  • Replace worn out or damaged parts
TRUSTED_SUSTAINABILITY
Specifications
Color Silver
For Use With (Equipment) Speedglas Welding Helmets 9100/9100 Air and 9100-QR
Product Type Heat Reflective Front