Learn More
3M™ Speedglas™ Ear and Neck Protection
Product Code. 15790088 Shop All 3M Products
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15790088 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15790088 Supplier 3M™ Supplier No. 7100080224
  • ONLINE EXCLUSIVE PRICE
    92.00€
    46.91€ / Pack of 3
    Save 45.09€
    49% Off
Estimated Shipment: 20-01-2027
3M™ Speedglas™ Ear and Neck Protection is designed for use with 3M™ Speedglas™ Welding Helmets 100, SL, 9100-QR, 9000 and 3M™ Welding Helmet 10.
  • For use with the complete helmet assembly to help protect the user from welding spark, spatter, and radiation on the ears and neck
TRUSTED_SUSTAINABILITY
Specifications
Color Black
For Use With (Equipment) 3M Speedglas Welding Helmets 100, SL, 9100-QR, 9000 and 3M Welding Helmet 10
Material Leather
Product Type Ear and Neck Protection