Learn More
Spectrum™ Spectra/Mesh Woven Polymer Macrofiltration Squares: N (Nylon)
Product Code. 15370912
Ansicht ändern
Click to view available options
Open Area:
14%
2%
21%
33%
36%
45%
47%
50%
55%
Size (Metric) Opening:
10μm
20μm
30μm
41μm
53μm
60μm
70μm
100μm
200μm
300μm
Packungsgröße:
Pack of 3
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Open Area Size (Metric) Opening unitSize
15370912 50% 300μm Pack of 3
11501150 55% 200μm Pack of 3
15380912 45% 60μm Pack of 3
15390912 36% 53μm Pack of 3
11757075 36% 70μm Pack of 3
11822512 33% 41μm Pack of 3
12654633 21% 30μm Pack of 3
11511150 14% 20μm Pack of 3
12568498 2% 10μm Pack of 3
10762926 47% 100μm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15370912 Lieferant Spectrum™ Lieferanten-Nr. 146486
Estimated Shipment: 07-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Combines high tensile strength with flexibility
Ideal for straining, filtering and screening solutions or suspensions containing high-molecular-weight molecules or particles.
  • Nonabsorbent
  • Combines high tensile strength with flexibility
Spezifikation
Adsorption High
Certifications/Compliance ISO 9001:2008
Material Nylon
Sterility Sterile
Hydrophobicity Hydrophilic
Open Area 50%
pH Range 3 to 10
Size (Metric) Opening 300μm
Temperature Stability 180°C
Thickness (Metric) 200 μm
Type Spectra Mesh Woven Filter
Diameter (English) 4.65 in.2
Mehr anzeigen Weniger anzeigen