Learn More
Spectrum™ Labs 8 - 10 kD Biotech RC Dialysis Kits
Product Code. 15380782
Click to view available options
Diameter (Metric):
6.4 mm
Unit Size:
0.5m
Product Code. 15380782
Brand: Spectrum™ 133 264T
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Includes
  • 0.5m Biotech RC tubing with glycerine
  • Two universal tubing closures
  • Five opening picks
With tubing made from Biotech RC membrane that can tolerate concentrated-weak acids, bases, dilute-strong acids, bases, most alcohols and some mild or dilute organic solvents
Specifications
MWCO 8 to 10 kDa
Diameter (Metric) 6.4 mm
Length (Metric) 0.32 mL/cm vol./length
Quantity 0.5 m Tubing/Kit