Learn More
Spectrum™ Spectra/Por Biotech-Grade RC Dialysis Membrane Tubing
Product Code. 13542040
Change view
Click to view available options
Diameter (Metric):
6.4 mm
10 mm
MWCO:
20 kDa
3.5 to 5 kDa
8 to 10 kDa
Unit Size:
5m
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) MWCO Unit Size
13542040 10 mm 20 kDa Each
13523880 6.4 mm 3.5 to 5 kDa Each
13464870 10 mm 3.5 to 5 kDa Each
15370782 6.4 mm 8 to 10 kDa 5m
15390782 10 mm 8 to 10 kDa 5m
15310792 6.4 mm 20 kDa 5m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 13542040 Supplier Spectrum™ Supplier No. 133342
  • ONLINE EXCLUSIVE PRICE
    512.00€
    339.00€ / Pack of 1
    Save 173.00€
    33% Off
Estimated Shipment: 18-12-2026
A premium dialysis membrane that provides superior molecular selectivity than Standard RC and superior tolerance than Biotech CE
  • Crafted from a regeneration process that yields superior temperature tolerances and chemical compatibilities with the same high purity and selectivity as Biotech CE
  • Membrane can tolerate concentrated-weak acids and bases, dilute-strong acids and bases, most alcohols and some mild or dilute organic solvents
  • Rigidly controlled porosity for better selectivity and separations
  • Ultrapure, does not interfere with molecular activity and function
  • 3 MWCOs and 2 tubing flat-widths
  • Packaged dry with glycerin (soak 15 min. in water)
  • Use with pH 2 to 12 and temp 4 to 60°C
Specifications
Product Type Spectra/Por Dialysis Tubing
MWCO 20 kDa
Material Regenerated Cellulose
Diameter (Metric) 10 mm
Diameter (English) 0.39 in.
Length (English) 16.4 ft.
Length (Metric) 5 m
Formulation Packaged dry with glycerin (soak 15 min. in water)
Temperature Range (English) 39.2°F to 140°F
Temperature Range (Metric) 4°C to 60°C
Quantity Each
For Use With (Application) Frequent dialysis applications, desalting, purification, sample preparation