Learn More
Spectrum™ Spectra/Por Biotech-Grade RC Dialysis Membrane Tubing
Click to view available options
Diameter (Metric):
6.4 mm
10 mm
MWCO:
20 kDa
3.5 to 5 kDa
8 to 10 kDa
Unit Size:
5m
Each
Description
- Crafted from a regeneration process that yields superior temperature tolerances and chemical compatibilities with the same high purity and selectivity as Biotech CE
- Membrane can tolerate concentrated-weak acids and bases, dilute-strong acids and bases, most alcohols and some mild or dilute organic solvents
- Rigidly controlled porosity for better selectivity and separations
- Ultrapure, does not interfere with molecular activity and function
- 3 MWCOs and 2 tubing flat-widths
- Packaged dry with glycerin (soak 15 min. in water)
- Use with pH 2 to 12 and temp 4 to 60°C
Specifications
Specifications
| Product Type | Spectra/Por Dialysis Tubing |
| MWCO | 3.5 to 5 kDa |
| Material | Regenerated Cellulose |
| Diameter (Metric) | 10 mm |
| Diameter (English) | 0.39 in. |
| Length (English) | 16.4 ft. |
| Length (Metric) | 5 m |
| Formulation | Packaged dry with glycerin (soak 15 min. in water) |
| Temperature Range (English) | 39.2°F to 140°F |
| Temperature Range (Metric) | 4°C to 60°C |
| Show More |