Learn More
Spectrum™ Spectra/Por™ 1-4 Standard RC Dialysis Membrane in Flat Sheets
Product Code. 12370170
Click to view available options
MWCO:
12 to 14 kDa
3.5 kDa
6 to 8 kDa
Width (English):
4.2 in.
5.9 in.
7.8 in.
9.4 in.
Length (English):
4.2 in.
5.9 in.
7.8 in.
9.4 in.
Membrane:
Spectra/Por 1
Spectra/Por 2
Spectra/Por 3
Spectra/Por 4
Unit Size:
Pack of 25
Product Code. 12370170
Brand: Spectrum™ 132712
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Clear, flexible and sturdy Standard RC (regenerated cellulose) in single layer square sheets that can be used with a variety of dialysis equipment or can be cut into specific dimensions for attaching to support frames for customized diagnostic assays, equilibrium studies, desalting, protein purification, etc.
  • Flat-Sheets deal for use with dialysis equipment or cutting to size
  • Available in 3.5 kD, 6-8 kD and 12-14 kD MWCO's
  • Good chemical compatibility with dilute strong acids & bases, conc. weak acids and bases, many alcohols & some organics.
  • Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
  • 25 sheets/pkg
Do not tie knots in tubing; use closures
Specifications
Product Type Spectra/Por 4 Dialysis Membrane Flat Sheets
MWCO 12 to 14 kDa
Membrane Spectra/Por 4
Length (English) 5.9 in.
Length (Metric) 150 mm
Width (English) 5.9 in.
Width (Metric) 150 mm
Dimensions (L x W) 5.9 x 5.9 in. (150 x 150 mm)
Color Clear
Temperature Range (English) 39°F to 250°F
Temperature Range (Metric) 4°C to 121°C
Quantity 25 Sheets
For Use With (Application) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting