Learn More
Biochrom Spectophotometer Calibration Set
Product Code. 15871192
Click to view available options
Type:
Spectophotometer Calibration Set
Unit Size:
Each
Product Code. 15871192
Brand: Biochrom 80211513
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Spectrophotometer Calibration Set used with Ultrospec 2100/7000/8000/9000, Libra S11/12/21/22/50/60/70/80, and BioDrop Spectrophotometers
NIST Calibrated with easy to use software
Specifications
Description Spectophotometer Calibration Set
Type Spectophotometer Calibration Set
For Use With (Equipment) Biochrom™ Ultrospec 2100/7000/8000/9000, Libra S11/12/21/22/50/60/70/80, and BioDrop Spectrophotometers