Learn More
VEE GEE Specific Gravity Hydrometer, 330 mm
Product Code. p-7250821
Click to view available options
Specific Gravity Range:
0.700 to 0.770
0.760 to 0.830
0.820 to 0.890
0.880 to 0.950
0.940 to 1.010
1.000 to 1.070
1.060 to 1.130
1.120 to 1.190
1.180 to 1.250
1.240 to 1.310
1.300 to 1.370
1.360 to 1.430
1.420 to 1.490
1.480 to 1.550
1.720 to 1.790
Unit Size:
Each
Product Code. 17836523
Brand: VEE GEE 660213
Only null left
Add to basket
Add to basket
The Ultra-Precision Specific Gravity Hydrometer features smaller ranges and higher resolution, with subdivisions of 0.0005. It is calibrated for use at 60°F.
  • Subdivisions: 0.0005
  • Approx. Length: 330 mm
Specifications
Type Specific Gravity Hydrometer
Increments 0.0005
Length (Metric) 33 cm
Material Glass
Specific Gravity Range 1.360 to 1.430
Temperature of Standardization 15.6°C (60°F)