Learn More
Greiner Bio-One Special Marker Pen
Click to view available options
Color:
Green
Tip Style:
Medium
Unit Size:
Pack of 10
Description
- Fine tip is excellent for general marking
- Highly resistant to abrasion and water
- Will not smear when subjected to water or alcohol during normal use
Specifications
Specifications
| Tip Style | Medium |
| Product Type | Marker Pen |
| Color | Green |
| Material | Polyethylene |