Learn More
SPE accessory filtration columns with frits polypropylene 6mL Bakerbond
Brand:  JT Baker 7121-06
133.00 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Additional Details : Weight : 0.19000kg
Product Code. 10076180
  • 140.00€ / Pack of 30
Estimated Shipment: 14-11-2024