Learn More
Brady™ SPC™ MRO15-DPS-E Absorbents MRO Plus Perforated Absorbent Roll
Description
MRO15-DPS-E is a 2-ply, grey, heavy weight, bonded and double perforated universal sorbent roll that absorbs industrial liquids including oils, water, solvents, coolants, etc.
- One sided spunbound gives extra strength as well as less lint.
- Perforated down the middle and cross-wise for ease of use and to minimize waste.
- A high quality universal sorbent for applications where a combination of good absorption and added strength is required.
- Ideal solution for catching overspray that comes from machinery.
Specifications
Specifications
| Color | Gray |
| Capacity (English) | 12.152 gal. |
| Capacity (Metric) | 46 L |
| Type | Absorbent Roll |
| Model | Universal Sorbent |
| Length (English) | 75.459 ft. |
| Length (Metric) | 23 m |
| Width (English) | 14.96 in. |
| Width (Metric) | 38 cm |
| Alternate Part Number | MRO15-DPS-E |