Learn More
Brady™ SPC™ Absorbents Universal Plus Chemical Boom
Click to view available options
Diameter (Metric):
13.00 cm
20.00 cm
Unit Size:
Pack of 4
Description
A chemical sorbent boom that is used to absorb and contain chemicals, hazardous liquids as well as oils and water-based fluids.
- Strong, durable outer sleeve keeps the absorbent material intact within the boom.
- Inner rope provides strength when deploying and retrieving the boom.
- Four staple clips at critical stress area add strength and prevent the boom from splitting during retrieval.
- Metal clips and rings allow linking of multiple booms.
Specifications
Specifications
| Color | Gray, Green |
| Capacity (English) | 68.95 gal. |
| Capacity (Metric) | 261 L |
| Type | Absorbent Boom |
| For Use With (Application) | Beverage Processing, Chemical, Food Processing, Laboratory, Manufacturing, Maritime, Oil and Gas, Pharmaceutical, Process, Warehouse and Logistics |
| Model | Chemical Sorbent |
| Length (English) | 9.843 ft. |
| Length (Metric) | 3 m |
| Diameter (English) | 7.874 in. |
| Diameter (Metric) | 20.00 cm |
| Show More |