missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ Absorbents Universal Plus Chemical Boom

Product Code. 30533337
Click to view available options
Diameter (Metric):
13.00 cm
20.00 cm
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 30533337

Brand: Brady™ 813849

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A chemical sorbent boom that is used to absorb and contain chemicals, hazardous liquids as well as oils and water-based fluids. Strong, durable outer sleeve keeps the absorbent material intact within the boom.

A chemical sorbent boom that is used to absorb and contain chemicals, hazardous liquids as well as oils and water-based fluids.

  • Strong, durable outer sleeve keeps the absorbent material intact within the boom.
  • Inner rope provides strength when deploying and retrieving the boom.
  • Four staple clips at critical stress area add strength and prevent the boom from splitting during retrieval.
  • Metal clips and rings allow linking of multiple booms.

Specifications

Color Gray, Green
Capacity (English) 31.436 gal.
Capacity (Metric) 119 L
Type Absorbent Boom
For Use With (Application) Beverage Processing, Chemical, Food Processing, Laboratory, Manufacturing, Maritime, Oil and Gas, Pharmaceutical, Process, Warehouse and Logistics
Model Chemical Sorbent
Length (English) 9.843 ft.
Length (Metric) 3 m
Diameter (English) 5.118 in.
Diameter (Metric) 13.00 cm
Alternate Part Number UN510
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.