Learn More
Brady™ SPC™ Absorbents Spill Trays
Product Code. 17891745 Shop All Brady Products
Change view
Click to view available options
Capacity (Metric):
20 L
30 L
40 L
60 L
Length (Metric):
40 cm
40.5 cm
80 cm
100 cm
Width (Metric):
60 cm
80.5 cm
Weight (Metric):
3 kg
4.5 kg
6.5 kg
9 kg
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Length (Metric) Width (Metric) Weight (Metric) Unit Size
17891745 40 L 80 cm 60 cm 6.5 kg Each
17801755 20 L 40 cm 60 cm 3 kg Each
17881745 30 L 40.5 cm 80.5 cm 4.5 kg Each
17821755 60 L 100 cm 60 cm 9 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17891745 Supplier Brady™ Supplier No. 306709
  • ONLINE EXCLUSIVE PRICE
    288.10€
    232.77€ / Pack of 1
    Save 55.33€
    19% Off
View all promotions
In Stock
Spill tray with black base and yellow platform.
  • For absorbents and spill control
Specifications
Color Yellow, Black
Capacity (English) 10.6 gal.
Capacity (Metric) 40 L
Material Polyethylene
Type Spill Tray
For Use With (Application) Beverage and Food Processing, Chemical, Healthcare, Manufacturing, Pharmaceutical, Laboratory
Length (English) 31.496 in.
Length (Metric) 80 cm
Width (English) 23.622 in.
Width (Metric) 60 cm
Height (English) 6.102 in.
Weight (English) 14.3 lb.
Weight (Metric) 6.5 kg
Alternate Part Number SPTY-40-YL-BK