missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ Absorbents Spill Containment Berm, QuickBerm Rigid Lock

Product Code. p-200113502
Click to view available options
Capacity (Metric) Sump:
662 L
890 L
2820 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30533386

Brand: Brady™ 198531

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The extreme durable Rigid-Lock QuickBerm offers immediate, secure deployment and spill-readiness. The built-in Rigid-Lock wall supports maximise usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter.

The extreme durable Rigid-Lock QuickBerm offers immediate, secure deployment and spill-readiness.

  • The built-in Rigid-Lock wall supports maximise usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter.
  • The high performance material withstands a broad range of chemicals including petroleum, oils, grease, and most acids and a broad UV resistance - durable enough for extended wear.

Specifications

Capacity (English) Sump 174.882 gal.
Capacity (Metric) Sump 662 L
Color Black
Material PVC-coated Fabric
Type Containment Berm
For Use With (Application) Beverage Processing, Chemical, Food Processing, Government, Healthcare, Manufacturing, Maritime, Mass transit, Oil and Gas, Pharmaceutical, Process, Warehouse and Logistics
Model Spill Containment
Length (English) 6.667 ft.
Length (Metric) 2.03 m
Width (English) 4.665 ft.
Width (Metric) 1.42 m
Height (English) 12.008 in.
Height (Metric) 30.50 cm
Alternate Part Number SB-QB-RL21
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.