missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ SPC™ Absorbents MAKE-A-BERM, Set of 2 Corners
Click to view available options
:
SB-MAB-COR Alternate Part Number
Description
SB-MAB-COR are 2 corners for the Make-A-Berm Spill Barrier.
- Made from durable vinyl and foam filled and resistant to oils, coolants and most chemicals.
- Allow you to make a barrier any shape you want.
Specifications
Specifications
| Color | Yellow |
| Material | Vinyl |
| Type | Containment Berm Kit |
| For Use With (Application) | Beverage Processing, Chemical, Food Processing, Government, Healthcare, Manufacturing, Maritime, Mass transit, Oil and Gas, Pharmaceutical, Process, Warehouse and Logistics |
| Model | Spill Containment |
| Alternate Part Number | SB-MAB-COR |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction