Learn More
Bochem™ Spatula, Cement
Product Code. 10020931
Click to view available options
Shape:
Pointed End
Round End
Round End, Pointed End
Unit Size:
Each
Product Code. 10020931
Brand: Bochem™ 3071
Spatula for fillings, cements and pastes. Bochem™ Spatula, Cement has a sturdy, ridged handle.
  • Stainless steel construction.
  • Sturdy, ridged handle.
Specifications
Shape Pointed End
For Use With (Application) Cements, fillings, pastes
Length (Metric) Blade 35 mm
Width (Metric) Blade 6 mm
Material Stainless Steel
Length (Metric) 150 mm