Learn More
Sparfloxacin, TRC
Specifications
Specifications
| Chemical Name or Material | Sparfloxacin |
| CAS | 110871-86-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H22 F2 N4 O3 |
| Quantity | 500 mg |
| Synonym | Sparfloxacin, 5-Amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethylpiperazin-1-yl]-6,8-difluoro-4-oxo-quinoline-3-carboxylic acid, 3-Quinolinecarboxylic acid','5-amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethyl-1-piperazinyl]-6,8-difluoro-1,4-dihydro-4-oxo-','rel-, 3-Quinolinecarboxylic acid','5-amino-1-cyclopropyl-7-(3,5-dimethyl-1-piperazinyl)-6,8-difluoro-1,4-dihydro-4-oxo-','cis-, rel-5-Amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethyl-1-piperazinyl]-6,8-difluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid, AT 4140, CI 978, CP 103826, PD 1315-1, PD 131501, Parox, Salocin 120, Spara, Sparcin, Zagam |
| InChI Formula | InChI=1S/C19H22F2N4O3/c1-8-5-24(6-9(2)23-8)17-13(20)15(22)12-16(14(17)21)25(10-3-4-10)7-11(18(12)26)19(27)28/h7-10,23H,3-6,22H2,1-2H3,(H,27,28)/t8-,9+ |
| SMILES | C[C@@H]1CN(C[C@H](C)N1)c2c(F)c(N)c3C(=O)C(=CN(C4CC4)c3c2F)C(=O)O |
| IUPAC Name | 5-amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethylpiperazin-1-yl]-6,8-difluoro-4-oxoquinoline-3-carboxylic acid |
| Molecular Weight (g/mol) | 392.4 |
| Show More |