missing translation for 'onlineSavingsMsg'
Learn More

Sparfloxacin, 98%

Product Code. 15497679
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497679 1 g 1g
15407689 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497679 Supplier Thermo Scientific Alfa Aesar Supplier No. J66358.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110871-86-8 | C19H22F2N4O3 | 392.41 g/mol

Targets a wide range of Gram-positive, Gram-negative, and mycoplasma species

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110871-86-8
Molecular Formula C19H22F2N4O3
Molecular Weight (g/mol) 392.41
MDL Number MFCD00869619
InChI Key DZZWHBIBMUVIIW-DTORHVGOSA-N
Synonym sparfloxacin, zagam, esparfloxacino, sparfloxacine, sparfloxacinum, spara, sparfloxacine inn-french, sparfloxacinum inn-latin, spfx, esparfloxacino inn-spanish
PubChem CID 60464
ChEBI CHEBI:9212
IUPAC Name 5-amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethylpiperazin-1-yl]-6,8-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
SMILES C[C@H]1CN(C[C@@H](C)N1)C1=C(F)C(N)=C2C(=O)C(=CN(C3CC3)C2=C1F)C(O)=O

Specifications

Melting Point 265°C
Boiling Point 640°C
Flash Point >110°C (230°F)
Packaging Plastic bottle
Quantity 1 g
Beilstein 9170271
Solubility Information Soluble in DMSO at 9mg/ml. Sparingly soluble in water
Formula Weight 392.4
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sparfloxacin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.