missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aqualytic™ SPADNS Reagent Solution
Click to view available options
Quantity:
10 mL
Unit Size:
250mL
Specifications
Specifications
| Packaging | 1 Each |
| Quantity | 10 mL |
Safety and Handling
Product Identifier
- SPADNS Reagent
- MIXTURE LIST-Contains : hydrochloric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction