Learn More
Honeywell Safety Products™ SP1000 2G Safety Eyewear
16.11 EUR valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Use promo code "21638" to get your promotional price.
Click to view available options
Lens Type: Impact Resistant
Description
- Provides protection against impact, wind, dust and airborne debris.
- Ideal for dirty, high-particulate work environments such as grinding, chipping, shaving and sanding.
- Soft and flexible nose piece.
- Conforms to a variety of facial profiles and minimizes slippage.
- Soft, high-quality face cushioning and padded temple tips.
- Comes with a wide headband.
- High-performance Honeywell™ K&N lens coatings for anti-fogging and scratch-resistance.
Specifications
Certifications/Compliance | EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number C9119.6HON, EC Attestation Number C9119.6HON Certifications• EN 166-2001 |
Lens Coating | Anti-fog, Impact-resistant |
Product Type | Safety glasses |
For Use With (Application) | Building, Construction, Metal, Steel, Mining, Quarrying, Petrochemical, and Transportation industries. |
Frame Color | Translucent Blue |
No. per Pack | 1 Ea. |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |