Learn More
Bel-Art™ Vibrasorb™ Premium Vibration Damping Mounts
Shop All SP Scienceware Products
missing translation for 'orderingAttributeHoverText'
Width (English):
35 x 35 x 7.6 cm
45 x 56 x 7.6 cm
missing translation for 'unitSize'
Each
Description
Protects electromechanical and optical equipment from exposure to vibration. Bel Art™ SP Scienceware™ Vibrasorb™ Premium Vibration Damping Mounts facilitates optimal equipment use.
- Polished surface resists scratches and chemical attack 6.4cm (2½ in.) thick platforms
- Molded of dense black & white terrazzo
- Four Vibro-Absorber™ cushions with neoprene feet support up to 16kg (35 lb)
- Larger size will absorb vibrations down to 13 Hz with a 16kg (35lb) load; smaller size will absorb vibrations down to 16 Hz. with a 16kg (35lb) load
- With lighter loads, the minimum frequency of damping for both sizes will increase to a value between 16 Hz. and 25 Hz
- Shipping weights: large 39kg (87lb), small 19kg (42lb)
Specifications
Specifications
| Product Type | Vibration Damping Mount |
| Color | Gray |
| Autoclavable | Not Autoclavable |
| Width (English) | 45 x 56 x 7.6 cm |
| Width (Metric) | 35 cm |
| Material | Terrazo |