missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Soybean Trypsin Inhibitor, ACF

Product Code. 17189271
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17189271 10 g 10g
17179271 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17189271 Supplier STEMCELL Technologies Supplier No. 07458

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For inhibition of trypsin activity

  • Soybean Trypsin Inhibitor, Animal Component-Free (ACF) is obtained from cultures free of animal-derived materials
  • This trypsin inhibitor consists of a single polypeptide chain with two disulfide bridges and inactivates trypsin on an equimolar basis
  • It is able to inhibit chymotrypsin to a lesser extent, but is ineffective against esterolytic, proteolytic, or elastolytic activities of porcine elastase (Vered et al.) Soybean Trypsin Inhibitor has also been shown to inhibit human and bovine thrombin, plasmin, and leukocytic proteases

Specifications

Synonym Antitrypsin; Trypsin inhibitor; Trypsin protease inhibitor
Cell Type Pancreatic Cells, T Cells, CD4+
Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.