missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Sotalol hydrochloride
GREENER_CHOICE

Product Code. p-7112851
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18035032

Brand: Tocris Bioscience™ 0952/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

β antagonist

Sotalol hydrochloride is a relatively potent pure β adrenergic antagonist, unique in possessing additional class III antiarrhythmic activity.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[4-[1-Hydroxy-2-[(1-methylethyl)amino]ethyl]phenyl]methanesulfonamide hydrochloride
CAS 959-24-0
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 50mM in water
Target Non-selective Adrenergic beta Receptor Antagonists
Molecular Formula C12H20N2O3S.HCl
Formula Weight Observed MW: 308.82
Purity 99%

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.