Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Buckets for 4-Place Swinging Bucket Rotors
Round or rectangular buckets for use with Four-Place Swinging Bucket Rotors 75006445 and 75006434
462.00€ - 550.00€
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10458191
|
Thermo Scientific™
75006441 |
Round Bucket; Capacity: 1 x 750 mL |
462.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
10638952
|
Thermo Scientific™
75006446 |
Rectangular Bucket, 1 x 250 mL |
550.00€
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
Description
Order Info
Sold individually; order correct quantity to equip rotor. Sealing lids sold separately. Accessory adapters accept assorted tube styles and volumes from 1.5 to 250mL.
Specifications
Swinging bucket rotor |