Learn More
Thermo Scientific™ Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Round Buckets
Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Tube Type:
15 mL DIN
7 mL Blood
7 mL DIN
Conical
Microtube
Tube Size:
1.5/2 mL
15 mL
50 mL
7 mL
missing translation for 'unitSize'
Each
Description
For use with four-place swinging bucket rotor 75-002-000 when equipped with round bucket 75-002-002
Adapters sold individually; order correct quantity to equip your rotor.
Specifications
Specifications
| Tube Type | Conical |
| For Use With (Equipment) | 4-place Swing rotor with round bucket |
| Tube Size | 15 mL |